Product Details
- SNP ID
-
rs12322796
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:132090744 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTGCATTACATGTATATGTTCAGTT[G/T]TGAAGGCATGACATTTTATGATAGT
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
EP400NL
PubMed Links
Gene Details
- Gene
- EP400NL
- Gene Name
- EP400 N-terminal like
There are no transcripts associated with this gene.
View Full Product Details