Product Details

SNP ID
rs12841891
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:18162959 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCATTTTAAATGTTTATTTATTG[G/T]ATCTATTTGGGAAACAAAACATTAA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BEND2 PubMed Links

Gene Details

Gene
BEND2
Gene Name
BEN domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184767.1 4604 Intron NP_001171696.1
NM_153346.4 4604 UTR 3 NP_699177.2
XM_011545447.1 4604 Intron XP_011543749.1
XM_017029266.1 4604 Intron XP_016884755.1
XM_017029267.1 4604 Intron XP_016884756.1

View Full Product Details