Product Details

SNP ID
rs12993324
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:73396225 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGATGTATTACATTAATTGATATTA[C/G]GTTATTAAACCAGCCTTGTGAAAAT
Phenotype
MIM: 606844
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ALMS1 PubMed Links

Gene Details

Gene
ALMS1
Gene Name
ALMS1, centrosome and basal body associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015120.4 Intron NP_055935.4

View Full Product Details