Product Details

SNP ID
rs4482466
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:68188588 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGCTGTACTAAAGGATTCTGTTGT[A/G]ACTCAGGCAGAGACATGAACTCTTC
Phenotype
MIM: 601302
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPP3R1 PubMed Links

Gene Details

Gene
PPP3R1
Gene Name
protein phosphatase 3 regulatory subunit B, alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000945.3 538 Missense Mutation TCA,TTA S49L NP_000936.1

View Full Product Details