Product Details

SNP ID
rs12234140
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32972486 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGCGGCGGCTCCCTAAACCACTTT[G/T]CGTGTTCATCCGCCTCCATCCGAGA
Phenotype
MIM: 601540
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BRD2 PubMed Links

Gene Details

Gene
BRD2
Gene Name
bromodomain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001113182.2 892 UTR 5 NP_001106653.1
NM_001199455.1 892 UTR 5 NP_001186384.1
NM_001199456.1 892 Intron NP_001186385.1
NM_001291986.1 892 UTR 5 NP_001278915.1
NM_005104.3 892 UTR 5 NP_005095.1

View Full Product Details