Product Details

SNP ID
rs12190988
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:52372026 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTCAGCACTCTATCTAAGAAAT[C/T]CTAGAAGCTGGGAATTTCTGGGGCT
Phenotype
MIM: 607780
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PAQR8 PubMed Links

Gene Details

Gene
PAQR8
Gene Name
progestin and adipoQ receptor family member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_133367.4 Intron NP_588608.1

View Full Product Details