Product Details

SNP ID
rs13362959
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:66595623 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTCACCAGGTTCTCTGAAAGTAG[A/G]GGAATAAACAAGGTTTTTGGATATA
Phenotype
MIM: 300276
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EDA2R PubMed Links

Gene Details

Gene
EDA2R
Gene Name
ectodysplasin A2 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199687.2 3546 Intron NP_001186616.1
NM_001242310.1 3546 Intron NP_001229239.1
NM_001324199.1 3546 Intron NP_001311128.1
NM_001324201.1 3546 Intron NP_001311130.1
NM_001324202.1 3546 Intron NP_001311131.1
NM_001324204.1 3546 Intron NP_001311133.1
NM_001324205.1 3546 Intron NP_001311134.1
NM_001324206.1 3546 Intron NP_001311135.1
NM_021783.4 3546 Intron NP_068555.1
XM_011531000.1 3546 UTR 3 XP_011529302.1
XM_011531001.1 3546 UTR 3 XP_011529303.1
XM_011531002.1 3546 UTR 3 XP_011529304.1
XM_011531003.2 3546 UTR 3 XP_011529305.1
XM_011531004.1 3546 Intron XP_011529306.1
XM_011531005.2 3546 UTR 3 XP_011529307.1
XM_011531006.2 3546 UTR 3 XP_011529308.1
XM_011531007.2 3546 Intron XP_011529309.1
XM_017029703.1 3546 UTR 3 XP_016885192.1
XM_017029704.1 3546 UTR 3 XP_016885193.1
XM_017029705.1 3546 UTR 3 XP_016885194.1
XM_017029706.1 3546 UTR 3 XP_016885195.1
XM_017029707.1 3546 UTR 3 XP_016885196.1
XM_017029708.1 3546 UTR 3 XP_016885197.1

View Full Product Details