Product Details

SNP ID
rs11944608
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:76117490 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCTATCTTTAATTTTTTGAGTAAA[C/T]ACTACACTGTTTTCCATAGAGGCTG
Phenotype
MIM: 603086 MIM: 607607
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ART3 PubMed Links

Gene Details

Gene
ART3
Gene Name
ADP-ribosyltransferase 3
There are no transcripts associated with this gene.

Gene
NUP54
Gene Name
nucleoporin 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278603.1 Intron NP_001265532.1
NM_017426.3 Intron NP_059122.2
XM_006714237.1 Intron XP_006714300.1
XM_011532033.2 Intron XP_011530335.1
XM_011532034.2 Intron XP_011530336.1
XM_017008294.1 Intron XP_016863783.1
XM_017008295.1 Intron XP_016863784.1

View Full Product Details