Product Details

SNP ID
rs11568853
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:109913569 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCATTTAACAAAGTTTGTCTTTGGG[C/T]ATGTATAGTTGCTTAATTTTTGTTT
Phenotype
MIM: 131530
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EGF PubMed Links

Gene Details

Gene
EGF
Gene Name
epidermal growth factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178130.2 Intron NP_001171601.1
NM_001178131.2 Intron NP_001171602.1
NM_001963.5 Intron NP_001954.2
XM_005262796.2 Intron XP_005262853.1
XM_011531707.1 Intron XP_011530009.1
XM_017007845.1 Intron XP_016863334.1
XM_017007846.1 Intron XP_016863335.1
XM_017007847.1 Intron XP_016863336.1
XM_017007848.1 Intron XP_016863337.1
XM_017007849.1 Intron XP_016863338.1
XM_017007850.1 Intron XP_016863339.1
XM_017007851.1 Intron XP_016863340.1
XM_017007852.1 Intron XP_016863341.1
XM_017007853.1 Intron XP_016863342.1
XM_017007854.1 Intron XP_016863343.1
XM_017007855.1 Intron XP_016863344.1

View Full Product Details