Product Details

SNP ID
rs12023262
Assay Type
Functionally tested
NCBI dbSNP Submissions
40
Location
Chr.1:42527060 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTTCATCATTTTTCAAAATAACT[A/G]CTCTGAAGGGTGTAAAGCTGACCAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC30 PubMed Links

Gene Details

Gene
CCDC30
Gene Name
coiled-coil domain containing 30
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080850.2 Intron NP_001074319.1
XM_011542067.2 Intron XP_011540369.1
XM_011542068.2 Intron XP_011540370.1
XM_011542069.2 Intron XP_011540371.1
XM_011542070.2 Intron XP_011540372.1
XM_011542071.2 Intron XP_011540373.1
XM_017002225.1 Intron XP_016857714.1
XM_017002226.1 Intron XP_016857715.1

View Full Product Details