Product Details

SNP ID
rs12134547
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:32474778 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAATATGGAGAAACTGTTACCTA[A/G]AAGGGTTTAGTGAGTTTCGTAAGAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZBTB8B PubMed Links

Gene Details

Gene
ZBTB8B
Gene Name
zinc finger and BTB domain containing 8B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145720.1 Intron NP_001139192.1

View Full Product Details