Product Details

SNP ID
rs12527165
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:167273083 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCTTTGTGACTGCCCAGGCAAGCC[A/G]CTAACGTGAACGTGCTCCTCTTACT
Phenotype
MIM: 607995
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UNC93A PubMed Links

Gene Details

Gene
UNC93A
Gene Name
unc-93 homolog A (C. elegans)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143947.1 Intron NP_001137419.1
NM_018974.3 Intron NP_061847.2
XM_011535905.2 Intron XP_011534207.1
XM_011535906.2 Intron XP_011534208.1
XM_011535907.2 Intron XP_011534209.1
XM_011535908.2 Intron XP_011534210.1
XM_017010958.1 Intron XP_016866447.1

View Full Product Details