Product Details

SNP ID
rs11749403
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:32690738 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTTCCCATATTGGCTGTGGTCTA[A/G]GCCATATGACATGCTGTGGCAAAGG
Phenotype
MIM: 108962
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NPR3 PubMed Links

Gene Details

Gene
NPR3
Gene Name
natriuretic peptide receptor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000908.3 Intron NP_000899.1
NM_001204375.1 Intron NP_001191304.1
NM_001204376.1 Intron NP_001191305.1
XM_005248309.1 Intron XP_005248366.1
XM_011514047.1 Intron XP_011512349.1
XM_011514048.2 Intron XP_011512350.1
XM_011514049.2 Intron XP_011512351.1
XM_011514050.2 Intron XP_011512352.1
XM_017009492.1 Intron XP_016864981.1

View Full Product Details