Product Details
- SNP ID
-
hCV31896562
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:189441540 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCGTGGTTCGTTGTGGCGGCAGCGA[C/G]TTGAACTTTAGGAGAGCTGTGTTCT
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
WDR75
PubMed Links
Gene Details
- Gene
- WDR75
- Gene Name
- WD repeat domain 75
There are no transcripts associated with this gene.
View Full Product Details