Product Details

SNP ID
rs12474565
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:162268391 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGTTGTGAAAATATACCCTTAGCT[C/T]ATGGTCTAGGAAATAAAAATAATGA
Phenotype
MIM: 606951
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IFIH1 PubMed Links

Gene Details

Gene
IFIH1
Gene Name
interferon induced with helicase C domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022168.3 Intron NP_071451.2

View Full Product Details