Product Details

SNP ID
rs7215698
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:43941173 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACAGCAGCATGCGGGGACCCCCAC[C/T]CCGAGAAGGCCAGCGTGTCCTCTTT
Phenotype
MIM: 167780
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC102724183 PubMed Links

Gene Details

Gene
LOC102724183
Gene Name
Wnt family member 9-like
There are no transcripts associated with this gene.

Gene
LOC107546764
Gene Name
uncharacterized LOC107546764
There are no transcripts associated with this gene.

Gene
PPY
Gene Name
pancreatic polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319209.1 Intron NP_001306138.1
NM_002722.4 Intron NP_002713.1
XM_011524978.2 Intron XP_011523280.1

View Full Product Details