Product Details

SNP ID
rs12071463
Assay Type
Functionally Tested
NCBI dbSNP Submissions
17
Location
Chr.1:15621452 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGTCACCTAGGCAGTAGTGCGCTC[A/G]CAGGTCACTGTAGCCTCAACCTCCT
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
DDI2 PubMed Links

Gene Details

Gene
DDI2
Gene Name
DNA damage inducible 1 homolog 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032341.4 Intron NP_115717.3

View Full Product Details