Product Details

SNP ID
rs12058073
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:15460472 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCATCTGCCAACAAACAGGTCATCG[A/T]TCCCCACAGTTATAGGGAGCCCGAT
Phenotype
MIM: 609443
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CELA2A PubMed Links

Gene Details

Gene
CELA2A
Gene Name
chymotrypsin like elastase family member 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033440.2 Intron NP_254275.1

View Full Product Details