Product Details

SNP ID
rs11950492
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:58969404 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTCTCACTTTTCCTCAGTTTCTCC[A/G]GATGTCTCAAGCTCATTTATGCCTC
Phenotype
MIM: 600129
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PDE4D PubMed Links

Gene Details

Gene
PDE4D
Gene Name
phosphodiesterase 4D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001104631.1 7507 UTR 3 NP_001098101.1
NM_001165899.1 7507 UTR 3 NP_001159371.1
NM_001197218.1 7507 UTR 3 NP_001184147.1
NM_001197219.1 7507 UTR 3 NP_001184148.1
NM_001197220.1 7507 UTR 3 NP_001184149.1
NM_001197221.1 7507 UTR 3 NP_001184150.1
NM_001197222.1 7507 UTR 3 NP_001184151.1
NM_001197223.1 7507 UTR 3 NP_001184152.1
NM_006203.4 7507 UTR 3 NP_006194.2
XM_005248537.3 7507 UTR 3 XP_005248594.1
XM_005248538.4 7507 UTR 3 XP_005248595.1
XM_011543469.1 7507 UTR 3 XP_011541771.1
XM_011543470.2 7507 UTR 3 XP_011541772.1
XM_011543471.1 7507 UTR 3 XP_011541773.1
XM_011543473.1 7507 UTR 3 XP_011541775.1
XM_011543477.2 7507 UTR 3 XP_011541779.1
XM_017009565.1 7507 UTR 3 XP_016865054.1
XM_017009566.1 7507 UTR 3 XP_016865055.1
XM_017009567.1 7507 UTR 3 XP_016865056.1
XM_017009568.1 7507 UTR 3 XP_016865057.1
XM_017009569.1 7507 UTR 3 XP_016865058.1
XM_017009570.1 7507 UTR 3 XP_016865059.1
XM_017009571.1 7507 UTR 3 XP_016865060.1

View Full Product Details