Product Details

SNP ID
rs11074523
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:22815612 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGAGGTCTCCAGTAAGAAAGATGC[C/G]TTAGGGAGTCTCAATCTCAAACCCA
Phenotype
MIM: 604056
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HS3ST2 PubMed Links

Gene Details

Gene
HS3ST2
Gene Name
heparan sulfate-glucosamine 3-sulfotransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006043.1 Intron NP_006034.1
XM_011546001.2 Intron XP_011544303.1
XM_011546002.2 Intron XP_011544304.1
Gene
LOC107984851
Gene Name
collagen alpha-1(XVI) chain-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017023983.1 Intron XP_016879472.1

View Full Product Details