Product Details

SNP ID
rs12026409
Assay Type
Functionally tested
NCBI dbSNP Submissions
43
Location
Chr.1:119031914 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCTGTGAATCCAGATTTTAAGTTC[A/G]CATCCATTGGCTGAATTGATGATAT
Phenotype
MIM: 604733
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WARS2 PubMed Links

Gene Details

Gene
WARS2
Gene Name
tryptophanyl tRNA synthetase 2, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015836.3 1930 UTR 3 NP_056651.1
NM_201263.2 1930 UTR 3 NP_957715.1
XM_005270350.3 1930 UTR 3 XP_005270407.1
XM_011540494.2 1930 UTR 3 XP_011538796.1
XM_011540495.2 1930 UTR 3 XP_011538797.1
XM_017000038.1 1930 UTR 3 XP_016855527.1
XM_017000039.1 1930 UTR 3 XP_016855528.1
XM_017000040.1 1930 UTR 3 XP_016855529.1
XM_017000041.1 1930 UTR 3 XP_016855530.1
XM_017000042.1 1930 UTR 3 XP_016855531.1

View Full Product Details