Product Details

SNP ID
rs10973223
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:37121840 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCCCACTGAAATACTGTGCTCTGT[C/T]TGTCTTTTGTGACTTTTGTTTTACA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZCCHC7 PubMed Links

Gene Details

Gene
ZCCHC7
Gene Name
zinc finger CCHC-type containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289119.1 Intron NP_001276048.1
NM_001289120.1 Intron NP_001276049.1
NM_001289121.1 Intron NP_001276050.1
NM_032226.2 Intron NP_115602.2
XM_005251608.4 Intron XP_005251665.1
XM_005251612.2 Intron XP_005251669.1
XM_011518050.1 Intron XP_011516352.1
XM_011518051.2 Intron XP_011516353.1
XM_017015197.1 Intron XP_016870686.1
XM_017015198.1 Intron XP_016870687.1
XM_017015199.1 Intron XP_016870688.1
XM_017015200.1 Intron XP_016870689.1
XM_017015201.1 Intron XP_016870690.1

View Full Product Details