Product Details

SNP ID
rs12351132
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:35759256 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTCATAATCCTGGTGCCAGATTT[C/T]TAAATTATTACAGACAATAGATGAT
Phenotype
MIM: 612191 MIM: 108961 MIM: 615742
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MSMP PubMed Links

Gene Details

Gene
MSMP
Gene Name
microseminoprotein, prostate associated
There are no transcripts associated with this gene.

Gene
NPR2
Gene Name
natriuretic peptide receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003995.3 Intron NP_003986.2
XM_005251478.3 Intron XP_005251535.1
XM_005251479.4 Intron XP_005251536.1
XM_011517890.2 Intron XP_011516192.1
XM_011517891.2 Intron XP_011516193.1
XM_011517892.2 Intron XP_011516194.1
XM_011517893.2 Intron XP_011516195.1
XM_011517895.2 Intron XP_011516197.1
XM_017014745.1 Intron XP_016870234.1
XM_017014746.1 Intron XP_016870235.1
XM_017014747.1 Intron XP_016870236.1
XM_017014748.1 Intron XP_016870237.1
XM_017014749.1 Intron XP_016870238.1
XM_017014750.1 Intron XP_016870239.1
Gene
RGP1
Gene Name
RGP1 homolog, RAB6A GEF complex partner 1
There are no transcripts associated with this gene.

View Full Product Details