Product Details

SNP ID
rs10967560
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:26845305 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGTTGGACTTCATGAGTTGTTCAT[C/T]TTCATCTCTTAACTTTTATTGCACA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CAAP1 PubMed Links

Gene Details

Gene
CAAP1
Gene Name
caspase activity and apoptosis inhibitor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167575.1 Intron NP_001161047.1
NM_024828.3 Intron NP_079104.3

View Full Product Details