Product Details

SNP ID
rs12338263
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:14717238 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTGTGGTGTTTTGTAGGCAAAAG[C/T]TTAGGGAATAAGACTCAGTATTGTG
Phenotype
MIM: 603777
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CER1 PubMed Links

Gene Details

Gene
CER1
Gene Name
cerberus 1, DAN family BMP antagonist
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005454.2 Intron NP_005445.1

View Full Product Details