Product Details

SNP ID
rs10975491
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:6215147 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTACTTCATCCCTTTATTTTCTG[C/G]TACAGATGCCAAACGAGATGGAGAG
Phenotype
MIM: 608678
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IL33 PubMed Links

Gene Details

Gene
IL33
Gene Name
interleukin 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199640.1 557 Intron NP_001186569.1
NM_001199641.1 557 Intron NP_001186570.1
NM_001314044.1 557 Intron NP_001300973.1
NM_001314045.1 557 Intron NP_001300974.1
NM_001314046.1 557 Intron NP_001300975.1
NM_001314047.1 557 Intron NP_001300976.1
NM_001314048.1 557 Intron NP_001300977.1
NM_033439.3 557 Intron NP_254274.1
XM_011518061.2 557 Intron XP_011516363.1
XM_017015285.1 557 UTR 5 XP_016870774.1

View Full Product Details