Product Details

SNP ID
rs10975044
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:5302095 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAATTTTCTATATACTTAAAGCCA[C/G]TTAAATGAGTCAGCCAAAACAGTTA
Phenotype
MIM: 179730 MIM: 179740
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RLN1 PubMed Links

Gene Details

Gene
RLN1
Gene Name
relaxin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006911.3 Intron NP_008842.1
Gene
RLN2
Gene Name
relaxin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005059.3 Intron NP_005050.2
NM_134441.2 Intron NP_604390.1
XM_017015011.1 Intron XP_016870500.1

View Full Product Details