Product Details
- SNP ID
-
rs10990414
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:102995847 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTAGTAAAAAGTGGGGCAGCTCTA[C/T]CCAAATGTGACCAACTGAATACTAG
- Phenotype
-
MIM: 604035
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CYLC2
PubMed Links
Gene Details
- Gene
- CYLC2
- Gene Name
- cylicin 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001340.4 |
|
Intron |
|
|
NP_001331.1 |
View Full Product Details