Product Details

SNP ID
rs7027778
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:111556631 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCCAAGGTTTAGGGTTTGTATCCA[A/G]TGCTGCGGGAGGGACCTGGTGGGAG
Phenotype
MIM: 601274
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PTGR1 PubMed Links

Gene Details

Gene
PTGR1
Gene Name
prostaglandin reductase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146108.1 Intron NP_001139580.1
NM_001146109.1 Intron NP_001139581.1
NM_012212.3 Intron NP_036344.2
XM_011518394.2 Intron XP_011516696.1
XM_011518395.2 Intron XP_011516697.1
XM_017014485.1 Intron XP_016869974.1
Gene
ZNF483
Gene Name
zinc finger protein 483
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007169.3 Intron NP_001007170.1
NM_133464.3 Intron NP_597721.2
XM_011518300.2 Intron XP_011516602.1
XM_017014337.1 Intron XP_016869826.1
XM_017014338.1 Intron XP_016869827.1
XM_017014339.1 Intron XP_016869828.1

View Full Product Details