Product Details
- SNP ID
-
rs11643021
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:71365394 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACATCTTGAGTGTGCTGAAGGGCTG[C/G]TTAGAACACAGGCTTCTGCACCCCA
- Phenotype
-
MIM: 114051
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CALB2
PubMed Links
Gene Details
- Gene
- CALB2
- Gene Name
- calbindin 2
View Full Product Details