Product Details

SNP ID
rs11913841
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:32362856 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCTCTTGATTAGGCAAATAAAAAT[C/T]TTGGCACATTTAAACATATGGCTTA
Phenotype
MIM: 605970 MIM: 605971
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RFPL3 PubMed Links

Gene Details

Gene
RFPL3
Gene Name
ret finger protein like 3
There are no transcripts associated with this gene.

Gene
RFPL3S
Gene Name
RFPL3 antisense
There are no transcripts associated with this gene.

Gene
SLC5A4
Gene Name
solute carrier family 5 member 4
There are no transcripts associated with this gene.

View Full Product Details