Product Details

SNP ID
rs11705335
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:38986392 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAAAATTATGCATTCCTGCACCG[C/G]ACGCTAAAGGAGATTCTCAGGTGAG
Phenotype
MIM: 607110
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
APOBEC3B PubMed Links

Gene Details

Gene
APOBEC3B
Gene Name
apolipoprotein B mRNA editing enzyme catalytic subunit 3B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270411.1 604 Silent Mutation CGC,CGG R183R NP_001257340.1
NM_004900.4 604 Silent Mutation CGC,CGG R183R NP_004891.4
Gene
APOBEC3B-AS1
Gene Name
APOBEC3B antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details