Product Details

SNP ID
rs11090094
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:42381929 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCCAGGGAGGCCTGCCTCTCACTT[A/C]GACTCCTCTGTACTGCCCCAGAAGC
Phenotype
MIM: 608740
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NFAM1 PubMed Links

Gene Details

Gene
NFAM1
Gene Name
NFAT activating protein with ITAM motif 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318323.1 4710 UTR 3 NP_001305252.1
NM_145912.6 4710 UTR 3 NP_666017.1
XM_005261374.4 4710 UTR 3 XP_005261431.1

View Full Product Details