Product Details
- SNP ID
-
rs12768993
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:93958280 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCACAACCCCTGTGTAAGGAAGGCC[A/G]TAAAGTCTGATGCACCCAGAGACAT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PIPSL
PubMed Links
Gene Details
- Gene
- PIPSL
- Gene Name
- PIP5K1A and PSMD4-like, pseudogene
There are no transcripts associated with this gene.
View Full Product Details