Product Details

SNP ID
rs7913668
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:93896519 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAACCTAAACAAATGCCTCTTTCA[A/G]TTATGCTAGTCATGCTCATCTCCAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC35G1 PubMed Links

Gene Details

Gene
SLC35G1
Gene Name
solute carrier family 35 member G1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134658.1 Intron NP_001128130.1
NM_153226.2 Intron NP_694958.1
XM_011539371.2 Intron XP_011537673.1
XM_011539373.2 Intron XP_011537675.1

View Full Product Details