Product Details
- SNP ID
-
rs11574791
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:159316662 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTGCACTGAGAGTGCAGGCCTGGG[C/G]TGGCCTTTGAGGGCCTGCTCACCTA
- Phenotype
-
MIM: 161561
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
IL12B
PubMed Links
Gene Details
- Gene
- IL12B
- Gene Name
- interleukin 12B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002187.2 |
|
Intron |
|
|
NP_002178.2 |
View Full Product Details