Product Details
- SNP ID
-
rs13155409
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:151277615 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGCGCTGCTGCTCACGGAGCCTAC[A/C]AGGGAGATGACCAAGTCCAGGCGGG
- Phenotype
-
MIM: 613109
MIM: 608332
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
GM2A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs56998614] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GM2A
- Gene Name
- GM2 ganglioside activator
There are no transcripts associated with this gene.
- Gene
- SLC36A3
- Gene Name
- solute carrier family 36 member 3
View Full Product Details