Product Details

SNP ID
rs8177425
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:151023312 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCCACCTTCTGACCCCCACTATCC[C/T]TTGACATGAGATTTCGTTCCAAGAG
Phenotype
MIM: 138321 MIM: 607714
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPX3 PubMed Links

Gene Details

Gene
GPX3
Gene Name
glutathione peroxidase 3
There are no transcripts associated with this gene.

Gene
TNIP1
Gene Name
TNFAIP3 interacting protein 1
There are no transcripts associated with this gene.

View Full Product Details