Product Details
- SNP ID
-
hCV31990998
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:75062218 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCATGTGATATATCCATGTGAAACA[C/T]TTGTAACAACAGTAAATATCCAACA
- Phenotype
-
MIM: 608704
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NMRK1
PubMed Links
Gene Details
- Gene
- NMRK1
- Gene Name
- nicotinamide riboside kinase 1
There are no transcripts associated with this gene.
View Full Product Details