Product Details

SNP ID
rs12340784
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:75094689 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAAAAAAAATGATACAGGGACAC[A/C]TTTCAGTGGGCTGATATGAGTGAGG
Phenotype
MIM: 608704 MIM: 610180
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NMRK1 PubMed Links

Gene Details

Gene
NMRK1
Gene Name
nicotinamide riboside kinase 1
There are no transcripts associated with this gene.

Gene
OSTF1
Gene Name
osteoclast stimulating factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012383.4 Intron NP_036515.4
XM_006717053.3 Intron XP_006717116.1
XM_006717055.3 Intron XP_006717118.1
XM_011518524.2 Intron XP_011516826.1
XM_011518525.2 Intron XP_011516827.1
XM_017014622.1 Intron XP_016870111.1

View Full Product Details