Product Details
- SNP ID
-
rs12353502
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:72524380 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAAAAACAGGATCAGTGTCTTTTTA[A/G]TATTTACCATAGTGCCTGATATGTA
- Phenotype
-
MIM: 606706
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TMC1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78897530] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TMC1
- Gene Name
- transmembrane channel like 1
View Full Product Details