Product Details

SNP ID
rs11873216
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:54718385 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAGCTGGGTTTGCAACTGAGTTTG[A/G]ATGGGAAAGAGATTTTGCTCAGGGC
Phenotype
MIM: 603869
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAB27B PubMed Links

Gene Details

Gene
RAB27B
Gene Name
RAB27B, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004163.4 Intron NP_004154.2
XM_005266733.1 Intron XP_005266790.1
XM_006722518.1 Intron XP_006722581.1
XM_011526131.1 Intron XP_011524433.1
XM_017025913.1 Intron XP_016881402.1
XM_017025914.1 Intron XP_016881403.1

View Full Product Details