Product Details

SNP ID
rs13248954
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:39597158 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTTATTATTGTTGAGTTTTAGGA[G/T]TTGTTTAGATATTTTGGTCCTTTAT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ADAM18 PubMed Links

Gene Details

Gene
ADAM18
Gene Name
ADAM metallopeptidase domain 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190956.1 Intron NP_001177885.1
NM_001320313.1 Intron NP_001307242.1
NM_014237.2 Intron NP_055052.1

View Full Product Details