Product Details

SNP ID
rs6557795
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:22092595 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTTTTTTTGAGACTCTGTCTCA[A/G]AAGAAAAAAAAAAAGTTGTCATTGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM160B2 PubMed Links

Gene Details

Gene
FAM160B2
Gene Name
family with sequence similarity 160 member B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022749.5 Intron NP_073586.5
XM_005273602.3 Intron XP_005273659.1
XM_006716381.2 Intron XP_006716444.1
XM_006716385.3 Intron XP_006716448.1
XM_017013732.1 Intron XP_016869221.1
XM_017013733.1 Intron XP_016869222.1
XM_017013734.1 Intron XP_016869223.1

View Full Product Details