Product Details
- SNP ID
-
rs13066173
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:42554347 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTTCTTTTTGCATAATCAGGTAAT[G/T]CTTTTTCATTTTTTTTAGGCAATGT
- Phenotype
-
MIM: 604028
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SEC22C
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114494769] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SEC22C
- Gene Name
- SEC22 homolog C, vesicle trafficking protein
View Full Product Details