Product Details

SNP ID
rs13319803
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:44593969 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCGTGGATAGATCTCCTTCCTCTG[A/G]TACAGAGAATAGGTGACATGTGCAA
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ZNF660 PubMed Links

Gene Details

Gene
ZNF660
Gene Name
zinc finger protein 660
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173658.2 Intron NP_775929.2
XM_005265090.4 Intron XP_005265147.1
XM_006713117.3 Intron XP_006713180.1
XM_011533622.2 Intron XP_011531924.1

View Full Product Details