Product Details

SNP ID
rs11846502
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:44930646 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCATTTCACACTTTAGATTTTTGAA[C/T]TTGGCATGATCAACCTGGTAAGCAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLHL28 PubMed Links

Gene Details

Gene
KLHL28
Gene Name
kelch like family member 28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308112.1 Intron NP_001295041.1
NM_017658.4 Intron NP_060128.2
XM_005267770.3 Intron XP_005267827.1
XM_006720174.3 Intron XP_006720237.1
XM_011536847.2 Intron XP_011535149.1
XM_011536849.2 Intron XP_011535151.1
XM_017021378.1 Intron XP_016876867.1

View Full Product Details