Product Details

SNP ID
rs12272341
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:102773870 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCTGCCAAAGGCTTCATCTCTGCT[A/G]CCCCTTCTAGAATATGAGCCCTTTT
Phenotype
MIM: 185260
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MMP10 PubMed Links

Gene Details

Gene
MMP10
Gene Name
matrix metallopeptidase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002425.2 Intron NP_002416.1
Gene
WTAPP1
Gene Name
Wilms tumor 1 associated protein pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details