Product Details

SNP ID
rs11224390
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:100693816 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTGAAGAGCTGATTTAGCAACTGT[C/T]CATTCATTTATTAACATTTCTTGGA
Phenotype
MIM: 615936
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP42 PubMed Links

Gene Details

Gene
ARHGAP42
Gene Name
Rho GTPase activating protein 42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152432.2 Intron NP_689645.2
XM_011542615.2 Intron XP_011540917.1
XM_011542616.2 Intron XP_011540918.1
XM_011542617.2 Intron XP_011540919.1
XM_017017237.1 Intron XP_016872726.1
XM_017017238.1 Intron XP_016872727.1
Gene
LOC100128386
Gene Name
uncharacterized LOC100128386
There are no transcripts associated with this gene.

View Full Product Details