Product Details

SNP ID
rs11601305
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:104995922 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTACACCATGAACCAGGAAATGC[C/T]TGAATGAAGAGAGCTTCCAGGGTTG
Phenotype
MIM: 602665
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CASP5 PubMed Links

Gene Details

Gene
CASP5
Gene Name
caspase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136109.1 Intron NP_001129581.1
NM_001136110.1 Intron NP_001129582.1
NM_001136112.1 Intron NP_001129584.1
NM_004347.3 Intron NP_004338.3
XM_011543021.2 Intron XP_011541323.1

View Full Product Details